Fmoc-Cys(tBu)-OH 98%
Lieferant: Thermo Fisher Scientific
Synonyme:
N-Fmoc-S-(tert-butyl)-L-cysteine, N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-S-(tert-butyl)-L-cysteine, N-Fmoc-S-tert-butyl-L-cysteine, Fmoc-Cys(tBu)-OH
431270050
431270250
ACRO431270050EA
57.5
EUR
ACRO431270050
ACRO431270250
Fmoc-Cys(tBu)-OH 98%
Fmoc-Cys(tBu)-OH
Formel:
C₂₂H₂₅NO₄S Molecular Weight: 399.505 g/mol Schmelzpunkt: 137.1…138.4 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00037130 CAS-Nummer: 67436-13-9 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 136 to 140 °C |
HPLC | ≥97.5 % |
Specific optical rotation | -21.0° to -23.0° (20 °C, 589 nm) (c=1, DMF) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...