trans-2-(4-Chlorophenyl)vinylboronic acid 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
(E)-(4-Chlorostyryl)boronic acid
431340010
431340100
ACRO431340010EA
113
EUR
ACRO431340010
ACRO431340100
trans-2-(4-Chlorophenyl)vinylboronic acid 97%
trans-2-(4-Chlorophenyl)vinylboronic acid
Formel:
C₈H₈BClO₂ Molecular Weight: 182,42 g/mol Schmelzpunkt: 133…134 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD02093767 CAS-Nummer: 154230-29-2 |
Spezifikation Testergebnisse
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
pH | 5.5 to 6.5 (10% aqueous suspension) |
Sodium oxide (Na2O) | ≤0.4 % |
Silicon dioxide (SiO2) | ≤0.03 % |
Iron(III)oxide (Fe2O3) | ≤0.03 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...