4-Amino-2-hydroxypyridine 97%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 4-Amino-2-pyridone, 4-Aminopyridin-2(1H)-one, 4-Amino-2(1H)-pyridinone, 4-Amino-2-pyridinol, 4-Amino-pyridin-2-ol, 4-amino-1,2-dihydropyridin-2-one, 3-Deazacytosine, 4-Amino-1H-pyridin-2-one

432080010 432080050
ACRO432080010EA 136 EUR
ACRO432080010 ACRO432080050
4-Amino-2-hydroxypyridine 97%
4-Amino-2-hydroxypyridine
Formel: C₅H₆N₂O
Molecular Weight: 110.12 g/mol
Schmelzpunkt: 206.5…208.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD02682913
CAS-Nummer: 38767-72-5
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to green to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.7 %
UV at 245 nm A: ≤0.046
Color scale ≤10 APHA
Residue after evaporation ≤0.0003 %
Acidity ≤0.0002 meq/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR