4-Brom-2-pyridon 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Bromopyridin-2-ol, 4-bromo-1,2-dihydropyridin-2-one, 4-Bromo-2-hydroxypyridine
432090010
432090050
ACRO432090010EA
154
EUR
ACRO432090010
ACRO432090050
4-Brom-2-pyridon 97%
4-Brom-2-pyridon
Formel:
C₅H₄BrNO Molecular Weight: 174,00 g/mol Schmelzpunkt: 203…208 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234041 CAS-Nummer: 36953-37-4 |
Spezifikation Testergebnisse
Appearance | White to light brown Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...