Methyl-3-amino-2-benzo[b]thiophencarboxylat 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
Methyl 3-aminobenzo[b]thiophene-2-carboxylate
432410010
432410050
ACRO432410010EA
39.9
EUR
ACRO432410010
ACRO432410050
Methyl-3-amino-2-benzo[b]thiophencarboxylat 97%
Methyl-3-amino-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₉NO₂S Molecular Weight: 207.25 g/mol Schmelzpunkt: 109…111 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00206744 CAS-Nummer: 35212-85-2 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 108 to 112 °C |
HPLC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...