5-Fluor-2-hydroxypyridin 97%

Lieferant: Thermo Scientific
Achtung

432580050 432580250
ACRO432580050EA 118 EUR
ACRO432580050 ACRO432580250
5-Fluor-2-hydroxypyridin 97%
5-Fluor-2-hydroxypyridin
Formel: C₅H₄FNO
Molecular Weight: 113,09 g/mol
Schmelzpunkt: 152…153 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03092918
CAS-Nummer: 51173-05-8

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Crystalline powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR