6-Iod-1H-indazol 97%
Lieferant: Thermo Scientific
Synonyme:
6-Iodoindazole
432420010
432420050
ACRO432420010EA
153
EUR
ACRO432420010
ACRO432420050
6-Iod-1H-indazol 97%
6-Iodo-1H-indazole
Formel:
C₇H₅IN₂ Molecular Weight: 244,0300 g/mol Schmelzpunkt: 208…211 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04114695 CAS-Nummer: 261953-36-0 |
Spezifikation Testergebnisse
Appearance | Yellow to orange Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 206 to 211 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...