2-Methyl-4-(4-morpholinyl)aniline 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Methyl-4-(4-morpholinyl)benzenamine
432460010
432460050
ACRO432460010EA
127
EUR
ACRO432460010
ACRO432460050
2-Methyl-4-(4-morpholinyl)aniline 97%
2-Methyl-4-(4-morpholinyl)aniline
Formel:
C₁₁H₁₆N₂O Molecular Weight: 192,26 g/mol Schmelzpunkt: 91…92 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10686817 CAS-Nummer: 581-00-0 |
Spezifikation Testergebnisse
Appearance | Yellow to brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 89 to 93 °C |
HPLC | ≥96.0 % |
Clarity of solution | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...