3-Amino-2-pyridincarbonsäure 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 3-Amino-2-pyridinecarboxylic acid, 3-Aminopyridine-2-carboxylic acid, 2-Pyridinecarboxylic acid, 3-amino-

432520050 432520250
ACRO432520050EA 114 EUR
ACRO432520050 ACRO432520250
3-Amino-2-pyridincarbonsäure 97%
3-Aminopicolinic acid
Formel: C₆H₆N₂O₂
Molecular Weight: 138,126 g/mol
Schmelzpunkt: 200…201 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00090153
CAS-Nummer: 1462-86-8

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR