5-Amino-2-methylbenzothiazol 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Methylbenzo[d]thiazol-5-amine, 2-Methyl-1,3-benzothiazol-5-amine

432760010 432760050
ACRO432760010EA 34.7 EUR
ACRO432760010 ACRO432760050
5-Amino-2-methylbenzothiazol 95%
5-Amino-2-methylbenzothiazole
Formel: C₈H₈N₂S
Molecular Weight: 164,23 g/mol
Schmelzpunkt: 101…103,5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00226291
CAS-Nummer: 13382-43-9
UN: 2811
ADR: 6.1,III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Crystalline powder
Infrared spectrum Conforms
Melting point 100 to 105 °C
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR