3-Amino-6-(morpholin-4-yl)pyridazine 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Amino-6-morpholinopyridazine

433270010 433270050
ACRO433270010EA 139 EUR
ACRO433270010 ACRO433270050
3-Amino-6-(morpholin-4-yl)pyridazine 97%
3-Amino-6-(morpholin-4-yl)pyridazine
Formel: C₈H₁₂N₄O
Molecular Weight: 180.21 g/mol
Schmelzpunkt: 181…183 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD09743734
CAS-Nummer: 66346-91-6

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥98.0 %
Residue after evaporation ≤0.001 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR