3-Cyanphenylboronsäurepinakolester 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile, 3-Cyanobenzeneboronic acid pinacol ester
433790010
433790050
ACRO433790010EA
73.4
EUR
ACRO433790010
ACRO433790050
3-Cyanphenylboronsäurepinakolester 97%
3-Cyanphenylboronsäurepinakolester
Formel:
C₁₃H₁₆BNO₂ Molecular Weight: 229,09 g/mol Schmelzpunkt: 80…81 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD03789262 CAS-Nummer: 214360-46-0 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 78 to 82 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...