3-Brom-1-propylboronsäurepinakolester 97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
3-Bromopropylboronic acid pinacol ester, 2-(3-Bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
433930010
433930050
ACRO433930010EA
49
EUR
ACRO433930010
ACRO433930050
3-Brom-1-propylboronsäurepinakolester 97%
3-Brom-1-propylboronsäurepinakolester
Formel:
C₉H₁₈BBrO₂ Molecular Weight: 248.96 g/mol Siedepunkt: 61…64 °C (.4 mmHg) Dichte: 1.177 g/cm³ Lagertemperatur: Raumtemperatur |
MDL:
MFCD10567053 CAS-Nummer: 124215-44-7 |
Spezifikation Testergebnisse
Appearance | Clear colorless to yellow Liquid |
Infrared spectrum | Conforms |
GC | ≥96.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...