2-(Carbobenzoxyamino)-1-ethanol 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-(Cbz-amino)-1-ethanol, N-Z-Ethanolamine, Benzyl N-(2-Hydroxyethyl)carbamate, Benzyl (2-hydroxyethyl)carbamate, Z-Glycinol, N-(2-Hydroxyethyl)carbamic Acid Benzyl Ester
433950050
433950250
ACRO433950050EA
26.8
EUR
ACRO433950050
ACRO433950250
2-(Carbobenzoxyamino)-1-ethanol 98%
2-(Carbobenzoxyamino)-1-ethanol
Formel:
C₁₀H₁₃NO₃ Molecular Weight: 195,22 g/mol Siedepunkt: 170 °C (,5Torr) Schmelzpunkt: 61…64 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00191060 CAS-Nummer: 77987-49-6 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 60 to 65 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...