Wasser ACS

Lieferant: Thermo Fisher Scientific

434400010 434400050
ACRO434400010EA 45.4 EUR
ACRO434400010 ACRO434400050
Wasser ACS
Wasser
Formel: H₂O
Siedepunkt: 100 °C (760 mmHg)
Schmelzpunkt: 0 °C
Dichte: 1 g/cm³
Lagertemperatur: Raumtemperatur
MDL: MFCD00011332
CAS-Nummer: 7732-18-5

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless Liquid
Chloride (Cl) ≤0.4 ppm
Nitrate (NO3) ≤0.4 ppm
Phosphate (PO4) ≤1.0 ppm
Sulfate (SO4) ≤1.0 ppm
Silicate (as SiO2) ≤0.01 ppm
Heavy metals (as Pb) ≤0.01 ppm
KMnO4-reducing substance Passes test
Conductivity 2.0 x 10-6 ohm-1 cm-1


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR