2-Brom-3,3,3-trifluorpropen 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 2-Bromo-3,3,3-trifluoro-1-propene

434460010 434460050
ACRO434460010EA 26 EUR
ACRO434460010 ACRO434460050
2-Brom-3,3,3-trifluorpropen 97%
2-Bromo-3,3,3-trifluoropropene
Formel: C₃H₂BrF₃
Molecular Weight: 174,95 g/mol
Siedepunkt: 34 °C (760 mmHg)
Dichte: 1,65 g/cm³
Lagertemperatur: Kühlschrank
MDL: MFCD00077469
CAS-Nummer: 1514-82-5
ADR: 3,I

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.3520 to 1.3570 (20°C, 589 nm)
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR