(R)-2-Amino-7-hydroxytetralin 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: (7R)-7-Amino-5,6,7,8-tetrahydronaphthalen-2-ol, (3R)-3-aminotetralin-6-ol, (7R)-7-Amino-5,6,7,8-tetrahydro-2-naphthalenol

435580010 435580050
ACRO435580010EA 141 EUR
ACRO435580010 ACRO435580050
(R)-2-Amino-7-hydroxytetralin 97%
(R)-2-Amino-7-hydroxytetralin
Formel: C₁₀H₁₃NO
Molecular Weight: 163,22 g/mol
Schmelzpunkt: 143,4…146.3 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11617807
CAS-Nummer: 85951-61-7

Bestellen

Spezifikation Testergebnisse

Appearance White to grey Crystalline powder
Infrared spectrum Conforms
Melting point 143 to 151 °C
Specific optical rotation +85° to +95° (20 °C, 589 nm) (c=0.5, methanol)
Titration with HClO4 96.0 to 104.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR