(2-Chlorphenyl)acetylen 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1-Chloro-2-ethynylbenzene

434750010 434750050
ACRO434750010EA 67.8 EUR
ACRO434750010 ACRO434750050
(2-Chlorphenyl)acetylen 98%
(2-Chlorphenyl)acetylen
Formel: C₈H₅Cl
Molecular Weight: 136,58 g/mol
Siedepunkt: 82 °C (16hPa)
Flammpunkt: 67 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00269951
CAS-Nummer: 873-31-4

Bestellen

Spezifikation Testergebnisse

Appearance Clear colorless to yellow to orange Liquid
Infrared spectrum Conforms
GC ≥97.5 %
HPLC 97.5 %
Titration with HClO4 97.5 % (On dry substance)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR