(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
N-Boc-L-serine benzyl ester, Boc-Ser-OBzl, BOC-L-Serine benzyl ester
434840010
434840050
ACRO434840010EA
53.9
EUR
ACRO434840010
ACRO434840050
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate 98%
(S)-Benzyl-2-((tert-butoxycarbonyl)amino)-3-hydroxypropanoate
Formel:
C₁₅H₂₁NO₅ Molecular Weight: 295,3 g/mol Schmelzpunkt: 67…72 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00076983 CAS-Nummer: 59524-02-6 |
Spezifikation Testergebnisse
Appearance | White Powder |
Infrared spectrum | Conforms |
Melting point | 65 to 75 °C |
HPLC | ≥97.5 % |
Specific optical rotation | -14° to -18° (20 °C, 589 nm) (c=1, ethanol) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...