L(+)-Cystein Hydrochlorid, wasserfrei 98%
Lieferant: Thermo Scientific
Achtung
Synonyme: H-Cys-OH.HCl, (R)-(+)-Cystein Hydrochlorid, (R)-(+)-2-Amino-3-mercaptopropansäure Hydrochlorid
434850010 434850250 434851000
ACRO434850010EA 1412 EUR
ACRO434850010 ACRO434850250 ACRO434851000
L(+)-Cystein Hydrochlorid, wasserfrei 98%
L(+)-Cystein Hydrochlorid
Formel: HSCH₂CH(NH₂)COOH·HCl
Molecular Weight: 157,62 g/mol
Schmelzpunkt: 175…181 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00064553
CAS-Nummer: 52-89-1
Bestellen
Spezifikation Testergebnisse
Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 8.67 to 9.11 % N
Specific optical rotation +6.5° to +9.5° (20°C, 589 nm) c=5 in 1N HCl