3-Cyanophenylacetic acid 95%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
m-Cyanophenylacetic acid, 2-(3-cyanophenyl)acetic Acid, Benzeneacetic acid,3-cyano-
434890010
434890050
ACRO434890010EA
124
EUR
ACRO434890010
ACRO434890050
3-Cyanophenylacetic acid 95%
3-Cyanophenylacetic acid
Formel:
C₉H₇NO₂ Molecular Weight: 161.16 g/mol Schmelzpunkt: 117…118 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD06411079 CAS-Nummer: 1878-71-3 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
Melting point | 113 to 119 °C |
HPLC | ≥94.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...