N-Acetyl-D-galactosamine 98%

Lieferant: Thermo Scientific

Synonyme: 2-Acetamido-2-deoxy-D-galactose, D-N-Acetylgalactosamine, 2-Acetamino-2-deoxy-alpha-D-galactopyranose, GalNAc, N-Acetylchondrosamine

434920010 434920050
ACRO434920010EA 202 EUR
ACRO434920010 ACRO434920050
N-Acetyl-D-galactosamine 98%
N-Acetyl-D-galactosamine
Formel: C₈H₁₅NO₆
Molecular Weight: 221,21 g/mol
Schmelzpunkt: 158…166 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00135832
CAS-Nummer: 1811-31-0

Bestellen

Spezifikation Testergebnisse

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤7.0 %
Solubility (10 % in water) Clear colorless
HPLC ≥97.5 %
Specific optical rotation +80.0° to +90.0° (20°C, 589 nm) (c=1, water, after 5 hours)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR