1,3-Dicyclohexylbenzimidazolium chloride 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: 1,3-Dicyclohexyl-1H-benzo[d]imidazol-3-ium chloride, 1,3-Dicyclohexyl-1H-benzimidazol-3-ium chloride

435090010 435095000
ACRO435090010EA 116 EUR
ACRO435090010 ACRO435095000
1,3-Dicyclohexylbenzimidazolium chloride 95%
1,3-Dicyclohexylbenzimidazolium chloride
Formel: C₁₉H₂₇ClN₂
Molecular Weight: 318,89 g/mol
Schmelzpunkt: 209,9…211.1 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD08705248
CAS-Nummer: 1034449-15-4

Bestellen

Spezifikation Testergebnisse

Appearance White to grey to tan Powder
Infrared spectrum Conforms
Elemental Analysis 8.25 to 9.31 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR