1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
Lieferant: Thermo Fisher Scientific
Synonyme:
(S,S)-Et-DUPHOS-Rh, (+)-1,2-BIS((2S,5S)-2,5-DIETHYLPHOSPHOLANO)BENZENE(CYCLOOCTADIENE)RHODIUM(L)TETRAFLUOROBORATE, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate, (+)-1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate, 1,2-Bis[(2S,5S)-2,5-diethylphospholano]benzene(1,5-cyclooctadiene)rhodium(I) tetrafluoroborate
435191000
435192500
ACRO435191000EA
105
EUR
ACRO435191000
ACRO435192500
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate 97%
1,2-Bis((2S,5S)-2,5-diethylphospholano)benzene(cyclooctadiene)rhodium(I) tetrafluoroborate
Formel:
C₃₀H₄₆BF₄P₂Rh Molecular Weight: 658.34 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD01862465 CAS-Nummer: 213343-64-7 |
Spezifikation Testergebnisse
Appearance | Orange to red to brown Powder |
Infrared spectrum | Conforms |
NMR (1H-NMR) | Consistent with structure |
NMR (31P-NMR) | Consistent with structure |
Elemental Analysis | 52.4 to 56.8 % C |
Specific optical rotation | -40° to -50° (20°C, 589 nm) (c=1 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...