(S)-2-Amino-7-hydroxytetralin 95%

Lieferant: Thermo Scientific
Achtung

Synonyme: (7S)-7-amino-5,6,7,8-tetrahydronaphthalen-2-ol, (3S)-3alpha-Aminotetralin-6-ol, (7S)-7-amino-5,6,7,8-tetrahydro-2-naphthalenol

435590010 435590050
ACRO435590010EA 170 EUR
ACRO435590010 ACRO435590050
(S)-2-Amino-7-hydroxytetralin 95%
(S)-2-Amino-7-hydroxytetralin
Formel: C₁₀H₁₃NO
Molecular Weight: 163,22 g/mol
Schmelzpunkt: 146,7…150.2 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD11506007
CAS-Nummer: 85951-60-6

Bestellen

Spezifikation Testergebnisse

Appearance White to grey Crystalline Powder or crystals
Infrared spectrum Conforms
Melting point 144 to 151 °C
Specific optical rotation -85° to -95° (20 °C, 589 nm) (c=0.5, MeOH)
Titration with HClO4 94 to 106 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR