Tetrakis(decyl)ammoniumbromid 98%

Lieferant: Thermo Scientific

Synonyme: Tetrakis(decyl)ammonium bromide, IPC-TDA-Br, Tetra(decyl)ammonium bromide

435670100 435670500
ACRO435670100EA 201 EUR
ACRO435670100 ACRO435670500
Tetrakis(decyl)ammoniumbromid 98%
Tetrakis(decyl)ammoniumbromid
Formel: C₄₀H₈₄BrN
Molecular Weight: 659,0057 g/mol
Schmelzpunkt: 88 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00043166
CAS-Nummer: 14937-42-9

Bestellen

Spezifikation Testergebnisse

Appearance White to light beige Powder
Infrared spectrum Conforms
Melting point 86 to 90 °C
Titration Argentometric ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR