3-Amino-5-brom-1H-indazol 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 5-Bromo-1H-indazol-3-amine

435760010 435760050
ACRO435760010EA 66.4 EUR
ACRO435760010 ACRO435760050
3-Amino-5-brom-1H-indazol 95%
3-Amino-5-bromo-1H-indazole
Formel: C₇H₆BrN₃
Molecular Weight: 212.05 g/mol
Schmelzpunkt: 237…238 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03426696
CAS-Nummer: 61272-71-7
UN: 2811
ADR: 6.1 III

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 174 to 178 °C
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR