4-Bromoisoindoline hydrochloride 97%
Lieferant: Thermo Scientific
|
Achtung
|
435870010
435870050
ACRO435870010EA
96.9
EUR
ACRO435870010
ACRO435870050
4-Bromoisoindoline hydrochloride 97%
4-Bromoisoindoline hydrochloride
Formel:
C₈H₉BrClN Schmelzpunkt: 269…274 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD10700134 CAS-Nummer: 923590-95-8 |
Spezifikation Testergebnisse
Appearance | Off-white to yellow to pink to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Elemental Analysis | 5.73 to 6.21 % N |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...