5,5-Diphenylhydantoin sodium salt 99%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
Phenytoin sodium, Sodium 5,5-Diphenylhydantoin, Sodium 2,4-dioxo-5,5-diphenylimidazolidin-1-ide
436100250
436101000
ACRO436100250EA
35
EUR
ACRO436100250
ACRO436101000
5,5-Diphenylhydantoin sodium salt 99%
5,5-Diphenylhydantoin sodium salt
Formel:
C₁₅H₁₂N₂O₂ Molecular Weight: 274,25 g/mol Schmelzpunkt: 295…298 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00069674 CAS-Nummer: 630-93-3 |
Spezifikation Testergebnisse
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | ≤3.0 % |
Titration with NaOH | 98.5 to 100.5 % (On anhydrous substance) |
Color of solution | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...