Acetobromo-α-D-glucuronic acid methyl ester 98%

Lieferant: Thermo Scientific
Achtung

Synonyme: BROMOMETHYLTRIACETYL GLUCURONATE, Acetobromo-α-D-glucuronic Acid Methyl Ester, Methyl acetobromo-α-D-glucuronate, (2,3,4-Tri-O-acetyl-α-D-glucopyranosyl bromide)uronic acid methyl ester, (2R,3R,4S,5S,6S)-2-Bromo-6-(methoxycarbonyl)tetrahydro-2H-pyran-3,4,5-triyl triacetate

436290010 436290050
ACRO436290010EA 190 EUR
ACRO436290010 ACRO436290050
Acetobromo-α-D-glucuronic acid methyl ester 98%
Acetobromo-alpha-D-glucuronic acid methyl ester
Formel: C₁₃H₁₇BrO₉
Molecular Weight: 397,174 g/mol
Schmelzpunkt: 80…110 °C
Lagertemperatur: Gefrierschrank
MDL: MFCD00061613
CAS-Nummer: 21085-72-3

Bestellen

Spezifikation Testergebnisse

Appearance White to grey Crystalline powder or solid
Infrared spectrum Conforms
HPLC ≥97.5 % (ELSD)
Stabilizer 2 % CaCO3
Specific optical rotation +185° to +195° (20°C, 589 nm) (c=1, ethyl acetate)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR