Sulfamethoxypyridazin
Lieferant: Thermo Scientific
|
Gefahr
|
Synonyme:
4-Amino-N-(6-methoxypyridazin-3-yl)benzenesulphonamide
436470050
436470250
ACRO436470050EA
39.3
EUR
ACRO436470050
ACRO436470250
Sulfamethoxypyridazin
Sulfamethoxypyridazin
Formel:
C₁₁H₁₂N₄O₃S Molecular Weight: 280,3 g/mol Schmelzpunkt: 182…183 °C Lagertemperatur: Kühlschrank |
MDL:
MFCD00057372 CAS-Nummer: 80-35-3 |
Spezifikation Testergebnisse
Appearance | White to yellow Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 180 to 184 °C |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...