6-Chlorpicolinsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
6-Chloropyridine-2-carboxylic acid, 2-Chloropyridine-6-carboxylic acid
436660010
436660050
ACRO436660010EA
29.5
EUR
ACRO436660010
ACRO436660050
6-Chlorpicolinsäure 97%
6-Chlorpicolinsäure
Formel:
C₆H₄ClNO₂ Molecular Weight: 157,56 g/mol Schmelzpunkt: 162,6…162.7 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00155390 CAS-Nummer: 4684-94-0 |
Spezifikation Testergebnisse
Appearance | White to cream to tan Crystalline powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...