2-Brom-5-iodpyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

436850050 436850250
ACRO436850050EA 66.8 EUR
ACRO436850050 ACRO436850250
2-Brom-5-iodpyridin 97%
2-Brom-5-iodpyridin
Formel: C₅H₃BrIN
Schmelzpunkt: 123…123,6 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03095201
CAS-Nummer: 73290-22-9

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to light pink to light brown Crystalline powder or needles
Infrared spectrum Conforms
Melting point 121 to 125 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR