2-Aminoisonicotinsäure 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
2-Amino-4-pyridinecarboxylic acid, 2-Aminopyridine-4-carboxylic acid
437360010
437360050
ACRO437360010EA
45.1
EUR
ACRO437360010
ACRO437360050
2-Aminoisonicotinsäure 97%
2-Aminoisonicotinsäure
Formel:
C₆H₆N₂O₂ Molecular Weight: 138,13 g/mol Schmelzpunkt: 345,2…347.3 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00233714 CAS-Nummer: 13362-28-2 |
Spezifikation Testergebnisse
Appearance | Off-white to brown Powder |
Infrared spectrum | Conforms |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...