4-Amino-2-brompyridin 95%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: 2-Brom-4-pyridylamin, 2-Brompyridin-4-amin

437410010 437410050
ACRO437410010EA 25.4 EUR
ACRO437410010 ACRO437410050
4-Amino-2-brompyridin 95%
4-Amino-2-brompyridin
Formel: C₅H₅BrN₂
Molecular Weight: 173.01 g/mol
Schmelzpunkt: 98…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01646061
CAS-Nummer: 7598-35-8

Bestellen

Spezifikation Testergebnisse

Appearance White to orange to light brown Powder
Infrared spectrum Conforms
HPLC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR