4-Iodnicotinonitril 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
3-Cyano-4-iodopyridine
437450010
437450050
ACRO437450010EA
354
EUR
ACRO437450010
ACRO437450050
4-Iodnicotinonitril 97%
4-Iodnicotinonitril
Formel:
C₆H₃IN₂ Molecular Weight: 230,01 g/mol Schmelzpunkt: 114…115 °C Lagertemperatur: Gefrierschrank |
MDL:
MFCD09037453 CAS-Nummer: 490039-72-0 ADR: 6.1,III |
Spezifikation Testergebnisse
Appearance | Light brown Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 112 to 116 °C |
GC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...