N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-β-alanin 99%
Lieferant: Thermo Scientific
Synonyme:
N-Boc-beta-alanine, Boc-β-Ala-OH, BOC-beta-alanine
437650050
437650250
437651000
ACRO437650050EA
20.4
EUR
ACRO437650050
ACRO437650250
ACRO437651000
N-{[(2-Methyl-2-propanyl)oxy]carbonyl}-β-alanin 99%
Boc-b-Alanine
Formel:
C₈H₁₅NO₄ Molecular Weight: 189,2 g/mol Schmelzpunkt: 77,5…78.5 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00037291 CAS-Nummer: 3303-84-2 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder or solid |
Infrared spectrum | Conforms |
Melting point | 76 to 80 °C |
Elemental Analysis | 7.29 to 7.51 % N |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...