5,6-Diaminouracil sulphate 95%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
5,6-Diamino-4-hydroxy-2(1H)-pyrimidinone sulphate, 5,6-Diamino-2,4-pyrimidinediol sulphate, 5,6-Diamino-2,4-dihydroxypyrimidine sulphate
437660100
437660500
ACRO437660100EA
109
EUR
ACRO437660100
ACRO437660500
5,6-Diaminouracil sulphate 95%
5,6-Diaminouracil sulphate
Formel:
C₄H₆N₄O₂.H₂SO₄ Molecular Weight: 240,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00135845 CAS-Nummer: 42965-55-9 |
Spezifikation Testergebnisse
Appearance | White to light brown Powder |
Infrared spectrum | Conforms |
Elemental Analysis | 21.93 to 27.40 % (N) |
Learn more
![](/m-de.vwr.com/de_DE/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...