3-Methoxyanthranilsäure 98%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2-Amino-3-methoxybenzoesäure

437720010 437720050 437720250
ACRO437720010EA 32.2 EUR
ACRO437720010 ACRO437720050 ACRO437720250
3-Methoxyanthranilsäure 98%
3-Methoxyanthranilsäure
Formel: C₈H₉NO₃
Molecular Weight: 167.16 g/mol
Schmelzpunkt: 168…170.7 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00075178
CAS-Nummer: 3177-80-8

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow to brown Powder
Infrared spectrum Conforms
HPLC ≥97.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR