2-Amino-3-brom-5-chlorpyridin 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 3-Brom-5-chlorpyridin-2-amin, 3-Brom-5-chlor-2-pyridylamin

437910010 437910050
ACRO437910010EA 45.8 EUR
ACRO437910010 ACRO437910050
2-Amino-3-brom-5-chlorpyridin 97%
2-Amino-3-brom-5-chlorpyridin
Formel: C₅H₄BrClN₂
Molecular Weight: 207.46 g/mol
Schmelzpunkt: 82…85 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00955627
CAS-Nummer: 26163-03-1

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Crystalline powder
Infrared spectrum Conforms
Melting point 83 to 87 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR