5-Amino-2,3-dihydrobenzo[b]furan 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 2,3-Dihydrobenzofuran-5-amine, 5-Amino-2,3-dihydrobenzofuran, 2,3-dihydro-1-benzofuran-5-amine, 5-Benzofuranamine,2,3-dihydro-

438030010 438030050
ACRO438030010EA 129 EUR
ACRO438030010 ACRO438030050
5-Amino-2,3-dihydrobenzo[b]furan 97%
5-Amino-2,3-dihydrobenzo[b]furan
Formel: C₈H₉NO
Molecular Weight: 135.166 g/mol
Schmelzpunkt: 81.6…83.4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03617969
CAS-Nummer: 42933-43-7

Bestellen

Spezifikation Testergebnisse

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 80 to 84 °C
GC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR