4-Brom-3-(trifluormethyl)-1H-pyrazol 97%
Lieferant: Thermo Scientific
|
Achtung
|
Synonyme:
4-Brom-3-(trifluormethyl)-1H-pyrazol
438040010
438040050
ACRO438040010EA
106
EUR
ACRO438040010
ACRO438040050
4-Brom-3-(trifluormethyl)-1H-pyrazol 97%
4-Bromo-3-(trifluoromethyl)-1H-pyrazole
Formel:
C₄H₂BrF₃N₂ Molecular Weight: 214,97 g/mol Schmelzpunkt: 112…114 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD04969230 CAS-Nummer: 19968-17-3 |
Spezifikation Testergebnisse
Appearance | White Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 111 to 115 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...