Methyl-4-brom-3-methylbenzoat 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Methyl 4-bromo-3-methylbenzoate

438190050 438190250
ACRO438190050EA 53.7 EUR
ACRO438190050 ACRO438190250
Methyl-4-brom-3-methylbenzoat 95%
Methyl-4-brom-3-methylbenzoat
Formel: C₉H₉BrO₂
Molecular Weight: 229.07 g/mol
Schmelzpunkt: 41…42.5 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00673014
CAS-Nummer: 148547-19-7

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Low melting solid or liquid
Infrared spectrum Conforms
Melting point 38 to 44 °C
GC ≥94.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR