(Methylsulfonyl)acetonitril 97%
Lieferant: Thermo Fisher Scientific
|
Gefahr
|
Synonyme:
Methanesulphonylacetonitrile, Cyanomethylsulphonylmethane, 2-(Methylsulphonyl)acetonitrile
438420050
438420250
ACRO438420050EA
66
EUR
ACRO438420050
ACRO438420250
(Methylsulfonyl)acetonitril 97%
(Methylsulfonyl)acetonitril
Formel:
C₃H₅NO₂S Molecular Weight: 119.14 g/mol Schmelzpunkt: 83…86 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00014743 CAS-Nummer: 2274-42-2 UN: 3439 ADR: 6.1 III |
Spezifikation Testergebnisse
Appearance | White to brown Powder |
Infrared spectrum | Conforms |
Melting point | 81 to 86 °C |
Elemental Analysis | 11.29 to 12.23 % (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...