(1R,2R,3R,5S)-(-)-Isopinocampheylamine 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (1R,2R,3R,5S)-2,6,6-Trimethylbicyclo[3.1.1]heptan-3-amine, (-)-Isopinocampheylamine, (1R,2R,3R,5S)-3-Pinanamine

439540010 439540050
ACRO439540010EA 84.7 EUR
ACRO439540010 ACRO439540050
(1R,2R,3R,5S)-(-)-Isopinocampheylamine 95%
(1R,2R,3R,5S)-(-)-Isopinocampheylamine
Formel: C₁₀H₁₉N
Molecular Weight: 153.27 g/mol
Siedepunkt: 90 °C (18 mmHg)
Flammpunkt: 72 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00192238
CAS-Nummer: 69460-11-3

Bestellen

Spezifikation Testergebnisse

Appearance Colorless to yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
Specific optical rotation -38.0° to -44.0° (20°C, 589 nm) (Neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR