Sulphisoxazole 99%

Lieferant: Thermo Scientific
Achtung

Synonyme: 4-Amino-N-(3,4-dimethylisoxazol-5-yl)benzenesulphonamide

439670250 439671000
ACRO439670250EA 35.2 EUR
ACRO439670250 ACRO439671000
Sulphisoxazole 99%
Sulphisoxazole
Formel: C₁₁H₁₃N₃O₃S
Molecular Weight: 267,303 g/mol
Schmelzpunkt: 194 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00003150
CAS-Nummer: 127-69-5

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR