trans-4-Brom-β-nitrostyrol 99%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: (E)-1-Bromo-4-(2-nitrovinyl)benzene

440160010 440160050
ACRO440160010EA 33.2 EUR
ACRO440160010 ACRO440160050
trans-4-Brom-β-nitrostyrol 99%
trans-4-Brom-β-nitrostyrol
Formel: C₈H₆BrNO₂
Schmelzpunkt: 148…152 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191856
CAS-Nummer: 5153-71-9

Bestellen

Spezifikation Testergebnisse

Appearance Yellow to yellow-green Powder
Infrared spectrum Conforms
Melting point 147 to 153 °C
GC ≥98.5 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR