Nα-Fmoc-N-δ-trityl-L-glutamine 95%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Fmoc-Gln(Trt)-OH

440640050 440640250
ACRO440640050EA 22.9 EUR
ACRO440640050 ACRO440640250
Nα-Fmoc-N-δ-trityl-L-glutamine 95%
N-alpha-Fmoc-N-delta-trityl-L-glutamine
Formel: C₃₉H₃₄N₂O₅
Molecular Weight: 610.71 g/mol
Schmelzpunkt: 131…144 °C
Lagertemperatur: Kühlschrank
MDL: MFCD00077056
CAS-Nummer: 132327-80-1

Bestellen

Spezifikation Testergebnisse

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Specific optical rotation -12.5° to -15.5° (20°C, 589 nm) (c=1 in DMF)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR