O-Benzyl-L-serine 99%

Lieferant: Thermo Fisher Scientific

Synonyme: H-Ser(Bzl)-OH

440930050 440930250
ACRO440930050EA 105 EUR
ACRO440930050 ACRO440930250
O-Benzyl-L-serine 99%
O-Benzyl-L-serine
Formel: C₁₀H₁₃NO₃
Molecular Weight: 195.22 g/mol
Schmelzpunkt: 210…213 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065937
CAS-Nummer: 4726-96-9

Bestellen

Spezifikation Testergebnisse

Appearance White to off-white Powder
Infrared spectrum Conforms
Water ≤1.0 %
HPLC ≥98.5 %
TLC ≥98.5 %
Specific optical rotation +20.0° to +25.0° (20°C, 589 nm) (c=1 in 8:2 AcOH/1M HCl)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR