9-Methylcarbazole-3-boronic acid pinacol ester 97%

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: 9-Methyl-3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-9H-carbazole, N-Methylcarbazole-3-boronic acid pinacol ester

441440010 441440050
ACRO441440010EA 83.6 EUR
ACRO441440010 ACRO441440050
9-Methylcarbazole-3-boronic acid pinacol ester 97%
9-Methylcarbazole-3-boronic acid pinacol ester
Formel: C₁₉H₂₂BNO₂
Molecular Weight: 307.20 g/mol
Schmelzpunkt: 151.7…152.4 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD16294552
CAS-Nummer: 1217891-71-8

Bestellen

Spezifikation Testergebnisse

Appearance Light brown Powder
Infrared spectrum Conforms
Melting point 147 to 154 °C
HPLC ≥96.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR